missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoate 99.0+%, TCI America™

Catalog No. p-7118335
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
B007425ML 25 mL
B0074500ML 500 mL
2 options

Catalog No. B007425ML

Supplier: TCI America B007425ML

Only null left

Chemical Identifiers

CAS 93-58-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00008421
InChI Key QPJVMBTYPHYUOC-UHFFFAOYSA-N
Synonym methylbenzoate, benzoic acid, methyl ester, clorius, benzoic acid methyl ester, niobe oil, oil of niobe, methyl benzenecarboxylate, essence of niobe, oniobe oil, oxidate le
PubChem CID 7150
ChEBI CHEBI:72775
IUPAC Name methyl benzoate
SMILES COC(=O)C1=CC=CC=C1

Specifications

Melting Point -12°C
Color Colorless
Boiling Point 200°C
Quantity 25 mL
Formula Weight 136.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH3850000
TSCA Yes