missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzimidate Hydrochloride 98.0+%, TCI America™

Catalog No. b09935g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B09935G 5 g
B09931G 1 g
2 options

Catalog No. B09935G

Supplier: TCI America B09935G

Only null left

Chemical Identifiers

CAS 5873-90-5
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.624
MDL Number MFCD00012575
InChI Key HDJNHVNQRJMWSH-UHFFFAOYSA-N
Synonym methyl benzimidate hydrochloride, methyl benzimidate hcl, methyl benzenecarboximidoate hydrochloride, benzimidic acid methyl ester hydrochloride, methyl benzenecarboximidate hydrochloride, benzenecarboximidic acid methyl ester hydrochloride, benzinidic acid methyl ester hydrochloride, benzenecarboximidic acid, methyl ester, hydrochloride, methoxyphenylmethanimine, chloride, acmc-1axhh
PubChem CID 5743287
IUPAC Name methyl benzenecarboximidate;hydrochloride
SMILES COC(=N)C1=CC=CC=C1.Cl

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 171.62
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Benzimidate Hydrochloride [for Protein modification]
TSCA No
Recommended Storage Refrigerator