accessibility menu, dialog, popup

UserName

HClCH3ONH

CAS RN 5873-90-5

IUPAC Name: hydrogen methyl benzenecarboximidate chloride
Synonyms: hydrogen methyl benzenecarboximidate chloride
Molecular Weight (g/mol): 171.62
Molecular Formula: C8H10ClNO
InChi Key: HDJNHVNQRJMWSH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=N)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results