missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzimidate Hydrochloride 98.0+%, TCI America™

Catalog No. B09931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09931G 1 g
B09935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09931G Supplier TCI America Supplier No. B09931G

Chemical Identifiers

CAS 5873-90-5
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.624
MDL Number MFCD00012575
InChI Key HDJNHVNQRJMWSH-UHFFFAOYSA-N
Synonym methyl benzimidate hydrochloride, methyl benzimidate hcl, methyl benzenecarboximidoate hydrochloride, benzimidic acid methyl ester hydrochloride, methyl benzenecarboximidate hydrochloride, benzenecarboximidic acid methyl ester hydrochloride, benzinidic acid methyl ester hydrochloride, benzenecarboximidic acid, methyl ester, hydrochloride, methoxyphenylmethanimine, chloride, acmc-1axhh
PubChem CID 5743287
IUPAC Name methyl benzenecarboximidate;hydrochloride
SMILES COC(=N)C1=CC=CC=C1.Cl

Specifications

Melting Point 107°C
Color White
Quantity 1 g
Formula Weight 171.62
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Benzimidate Hydrochloride [for Protein modification]
TSCA No
Recommended Storage Refrigerator