missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzethonium chloride, MP Biomedicals™

Catalog No. ICN19024010
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19024010 10 g
ICN19024025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19024010 Supplier MP Biomedicals Inc Supplier No. 0219024010

Chemical Identifiers

CAS 25155-18-4
Molecular Formula C28H44ClNO2
Molecular Weight (g/mol) 462.115
InChI Key QWZLBLDNRUUYQI-UHFFFAOYSA-M
PubChem CID 5702238
IUPAC Name benzyl-dimethyl-[2-[2-[2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC1=C(C=CC(=C1)C(C)(C)CC(C)(C)C)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

Color White
Quantity 10 g
Synonym methylbenzethonium chloride, methyl benzethonium chloride, unii-nn7590iuqx, nn7590iuqx, benzyldimethyl 2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium chloride, quat20, quat 20, prestwick_9, metilbenzetonio cloruro, benzyl-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium;chloride
Formula Weight 462.1
Physical Form Powder
Chemical Name or Material Methyl Benzethonium chloride
EINECSNumber 246-675-7