missing translation for 'onlineSavingsMsg'
Learn More

Methyl Aziridine-2-carboxylate (stabilized with HQ) 97.0+%, TCI America™

Catalog No. A12075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12075G Supplier TCI America Supplier No. A12075G
Only null left

Chemical Identifiers

CAS 5950-34-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00143640
InChI Key ZWCVDRJTYFIPIV-UHFFFAOYSA-N
Synonym Aziridine-2-carboxylic Acid Methyl Ester, 2-Methoxycarbonylaziridine
PubChem CID 534940
IUPAC Name methyl aziridine-2-carboxylate
SMILES COC(=O)C1CN1

Specifications

Color Yellow
Boiling Point 143°C
Quantity 5 g
UN Number 1992
Formula Weight 101.11
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl Aziridine-2-carboxylate (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator