accessibility menu, dialog, popup

UserName

CAS RN 5950-34-5

CH3OOHN

CAS RN 5950-34-5

IUPAC Name: methyl aziridine-2-carboxylate
Synonyms: methyl aziridine-2-carboxylate
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: ZWCVDRJTYFIPIV-UHFFFAOYNA-N
SMILES: COC(=O)C1CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results