missing translation for 'onlineSavingsMsg'
Learn More

Methyl Arachidate, 99%

CAS: 1120-28-1 | C21H42O2 | 326.57 g/mol

$152.97 - $578.03

Chemical Identifiers

CAS 1120-28-1
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.57
MDL Number MFCD00009014
InChI Key QGBRLVONZXHAKJ-UHFFFAOYSA-N
Synonym methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
PubChem CID 14259
IUPAC Name methyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC452820010
View Documents Promotion Details
N/A 1 g
Each for $152.97
Only null left
 
AC452820050
View Documents Promotion Details
N/A 5 g
Each for $578.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1120-28-1
326.57
QGBRLVONZXHAKJ-UHFFFAOYSA-N
14259
CCCCCCCCCCCCCCCCCCCC(=O)OC
C21H42O2
MFCD00009014
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
methyl icosanoate

Specifications

1120-28-1
215.0°C to 216.0°C (10.0 mmHg)
98.5 % min. (GC)
C21H42O2
MFCD00009014
QGBRLVONZXHAKJ-UHFFFAOYSA-N
methyl icosanoate
14259
99%
45.0°C to 48.0°C
>110°C
Glass Bottle
CH3(CH2)18COOCH3
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
CCCCCCCCCCCCCCCCCCCC(=O)OC
326.57
326.56
Methyl arachidate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-304-8

SDS
Documents

Documents

RUO – Research Use Only