missing translation for 'onlineSavingsMsg'
Learn More

Methyl arachidate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187571
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012483 1 g
111012484 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012483 Supplier Merck Emd Millipore Supplier No. 109411G
Only null left

Specifications

CAS 1120-28-1
Flash Point >113°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)18 COOCH3
MDL Number MFCD00009014
Melting Point 45°C to 48°C; 46°C to 49°C
Boiling Point 215°C to 216°C (10 mmHg)
Synonym Arachidic acid methyl ester; Methyl eicosanoate
Type Analytical Standard
Quantity 1 g
Formula Weight 326.56
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less