missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl alpha-D-Mannopyranoside 98.0+%, TCI America™
$85.44 - $520.23
Chemical Identifiers
| CAS | 617-04-9 |
|---|---|
| Molecular Formula | C7H14O6 |
| Molecular Weight (g/mol) | 194.18 |
| MDL Number | MFCD00063262 |
| InChI Key | HOVAGTYPODGVJG-UHFFFAOYNA-N |
| Synonym | methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside |
| PubChem CID | 101798 |
| ChEBI | CHEBI:43943 |
| IUPAC Name | 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
| SMILES | COC1OC(CO)C(O)C(O)C1O |
Chemical Identifiers
| 617-04-9 | |
| 194.18 | |
| HOVAGTYPODGVJG-UHFFFAOYNA-N | |
| 101798 | |
| 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
| C7H14O6 | |
| MFCD00063262 | |
| methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
| CHEBI:43943 | |
| COC1OC(CO)C(O)C(O)C1O |
Specifications
| 617-04-9 | |
| White | |
| MFCD00063262 | |
| methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
| COC1OC(CO)C(O)C(O)C1O | |
| 194.18 | |
| CHEBI:43943 | |
| ≥98.0% (HPLC) | |
| Methyl alpha-D-Mannopyranoside |
| 192°C | |
| C7H14O6 | |
| 25 g | |
| HOVAGTYPODGVJG-UHFFFAOYNA-N | |
| 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
| 101798 | |
| 194.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No