missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-Mannopyranoside 98.0+%, TCI America™

Supplier:  TCI America M0368250G

 View more versions of this product

Catalog No. M0368250G


Only null left

Description

Description

Specifications

Specifications

Methyl alpha-D-Mannopyranoside
192°C
C7H14O6
250 g
HOVAGTYPODGVJG-UHFFFAOYNA-N
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
101798
194.18
Crystalline Powder
617-04-9
White
MFCD00063262
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
COC1OC(CO)C(O)C(O)C1O
194.18
CHEBI:43943
≥98.0% (HPLC)

Chemical Identifiers

617-04-9
194.18
HOVAGTYPODGVJG-UHFFFAOYNA-N
101798
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
C7H14O6
MFCD00063262
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
CHEBI:43943
COC1OC(CO)C(O)C(O)C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions