missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl alpha-D-Glucopyranoside 98.0+%, TCI America™
$47.46 - $193.66
Chemical Identifiers
| CAS | 97-30-3 |
|---|---|
| Molecular Formula | C7H14O6 |
| Molecular Weight (g/mol) | 194.183 |
| MDL Number | MFCD00064086 |
| InChI Key | HOVAGTYPODGVJG-IECVIRLLSA-N |
| Synonym | methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r |
| PubChem CID | 7568545 |
| IUPAC Name | (2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
| SMILES | COC1C(C(C(C(O1)CO)O)O)O |
Chemical Identifiers
| C7H14O6 | |
| MFCD00064086 | |
| methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r | |
| (2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
Specifications
| 97-30-3 | |
| White | |
| MFCD00064086 | |
| HOVAGTYPODGVJG-IECVIRLLSA-N | |
| (2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
| 7568545 | |
| ≥98.0% (GC) | |
| Methyl alpha-D-Glucopyranoside |
| 170°C | |
| C7H14O6 | |
| methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r | |
| COC1C(C(C(C(O1)CO)O)O)O | |
| 194.183 | |
| 194.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (8)-0051
TSCA : Yes