missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-Glucopyranoside 98.0+%, TCI America™

Supplier:  TCI America M0228100G

Quantity 100 g

 View more versions of this product

Catalog No. M0228100G


Only null left

Description

Description

Specifications

Chemical Identifiers

97-30-3
194.183
HOVAGTYPODGVJG-IECVIRLLSA-N
7568545
COC1C(C(C(C(O1)CO)O)O)O
C7H14O6
MFCD00064086
methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r
(2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol

Specifications

Methyl alpha-D-Glucopyranoside
170°C
C7H14O6
MFCD00064086
HOVAGTYPODGVJG-IECVIRLLSA-N
(2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
7568545
≥98.0% (GC)
97-30-3
White
100 g
methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r
COC1C(C(C(C(O1)CO)O)O)O
194.183
194.18
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0051

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions