missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-Galactopyranoside Monohydrate 98.0+%, TCI America™

Catalog No. M10475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10475G 5 g
M104725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10475G Supplier TCI America Supplier No. M10475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34004-14-3
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00150748
InChI Key HOVAGTYPODGVJG-PZRMXXKTSA-N
Synonym methyl alpha-d-galactopyranoside monohydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol hydrate, methyl-alpha-d-galactopyranoside, methyl ?-galactoside hydrate, methyl a-d-galactopyranoside,monohydrate, methyl alpha-d-galactopyranoside hydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxyoxane-3,4,5-triol hydrate
PubChem CID 2724703
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Formula Weight 194.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl alpha-D-Galactopyranoside Monohydrate