accessibility menu, dialog, popup

UserName

CAS RN 34004-14-3

IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms: methyl α-galactoside methyl α-D-galactoside methyl-α-D-galactose me-α-gal α-methyl D-galactopyranoside α-methylgalactoside α-methyl D-galactoside
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: HOVAGTYPODGVJG-PZRMXXKTSA-N
SMILES: CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results