missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl alpha-Bromophenylacetate 98.0+%, TCI America™
$130.48 - $365.35
Chemical Identifiers
| CAS | 3042-81-7 |
|---|---|
| Molecular Formula | C9H9BrO2 |
| Molecular Weight (g/mol) | 229.073 |
| MDL Number | MFCD00013535 |
| InChI Key | NHFBYYMNJUMVOT-UHFFFAOYSA-N |
| Synonym | methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate |
| PubChem CID | 137806 |
| IUPAC Name | methyl 2-bromo-2-phenylacetate |
| SMILES | COC(=O)C(C1=CC=CC=C1)Br |
Chemical Identifiers
| 3042-81-7 | |
| 229.073 | |
| NHFBYYMNJUMVOT-UHFFFAOYSA-N | |
| 137806 | |
| COC(=O)C(C1=CC=CC=C1)Br |
| C9H9BrO2 | |
| MFCD00013535 | |
| methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate | |
| methyl 2-bromo-2-phenylacetate |
Specifications
| 3042-81-7 | |
| 172°C | |
| MFCD00013535 | |
| 3265 | |
| NHFBYYMNJUMVOT-UHFFFAOYSA-N | |
| methyl 2-bromo-2-phenylacetate | |
| 137806 | |
| ≥98.0% (GC) | |
| Methyl alpha-Bromophenylacetate |
| Yellow | |
| C9H9BrO2 | |
| 5 g | |
| methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate | |
| COC(=O)C(C1=CC=CC=C1)Br | |
| 229.073 | |
| 229.07 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator