missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-Bromo-2-chlorophenylacetate 98.0+%, TCI America™

Catalog No. p-7133072
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21755G 5 g
M217525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21755G Supplier TCI America Supplier No. M21755G

Chemical Identifiers

CAS 85259-19-4
Molecular Formula C9H8BrClO2
Molecular Weight (g/mol) 263.52
MDL Number MFCD08444353
InChI Key HMBUCZUZRQQJQD-UHFFFAOYNA-N
Synonym alpha-Bromo-2-chlorophenylacetic Acid Methyl Ester
PubChem CID 10730282
IUPAC Name methyl 2-bromo-2-(2-chlorophenyl)acetate
SMILES COC(=O)C(Br)C1=CC=CC=C1Cl

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 3265
Formula Weight 263.52
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Methyl alpha-Bromo-2-chlorophenylacetate
TSCA No
Recommended Storage Refrigerator