missing translation for 'onlineSavingsMsg'
Learn More

Methyl Alcohol, NF, 99.5%, Spectrum™ Chemical

ME151, 67-56-1, CH3OH

$270.07 - $5725.57

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604413
View Documents
Spectrum Chemical Mfg Cor
ME151500MLGL
500 mL Amber Glass Bottle
Each for $270.07
Only null left
 
18604414
View Documents
Spectrum Chemical Mfg Cor
ME151500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $1,166.90
Only null left
 
18604415
View Documents
Spectrum Chemical Mfg Cor
ME1514LTGL
4 L Amber Glass Bottle
Each for $683.30
Only null left
 
18604416
View Documents
Spectrum Chemical Mfg Cor
ME1514LTGLCS4
4 x 4 L Amber Glass Bottle
Case of 4 Each for $2,292.10
Only null left
 
18604417
View Documents
Spectrum Chemical Mfg Cor
ME15120LTMT
20 L Steel Can
Each for $1,839.83
Only null left
 
18604418
View Documents
Spectrum Chemical Mfg Cor
ME151200LTBL
200 L Drum
Each for $5,725.57
Only null left
 
Description

Description

Spectrum™ Chemical Methyl Alcohol, NF is as a solvent excipient for pharmaceutical products. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
CO
CH4O
MFCD00004595
methanol

Specifications

67-56-1
3 ppm
500 mL
CH4O
UN1230
CO
32.04
NF
100%
64°C to 65°C
Amber Glass Bottle
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
99.5%
Methyl Alcohol
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'