missing translation for 'onlineSavingsMsg'
Learn More

Methyl Alcohol, NF, 99.5%, Spectrum™ Chemical

ME151, 67-56-1, CH3OH

Supplier:  Spectrum Chemical Mfg Cor ME15120LTMT

 View more versions of this product

Catalog No. 18604417


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Methyl Alcohol, NF is as a solvent excipient for pharmaceutical products. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Methyl Alcohol
100%
64°C to 65°C
Steel Can
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
99.5%
67-56-1
3 ppm
20 L
CH4O
UN1230
CO
32.04
NF

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
CO
CH4O
MFCD00004595
methanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions