missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acrylate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. A0145500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0145500ML 500 mL
A014525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0145500ML Supplier TCI America Supplier No. A0145500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-33-3
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008627
InChI Key BAPJBEWLBFYGME-UHFFFAOYSA-N
Synonym methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
PubChem CID 7294
ChEBI CHEBI:82482
IUPAC Name methyl prop-2-enoate
SMILES COC(=O)C=C

Specifications

Melting Point -75°C
Color Colorless
Boiling Point 80°C
Quantity 500 mL
UN Number 1919
Formula Weight 86.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0987
RTECSNumber AT2800000
TSCA Yes