missing translation for 'onlineSavingsMsg'
Learn More

Methyl acrylate, 99%, stab. with ca 15ppm 4-methoxyphenol

Catalog No. AAA13128AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13128AE 100 mL
AAA13128AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA13128AE Supplier Thermo Scientific Chemicals Supplier No. A13128AE

CAS: 96-33-3 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl acrylate is a monomer used in the preparation of polymers having rigid, flexible, ionic, nonionic, hydrophobic, or hydrophilic properties. It is involved in the production of acrylate fiber, which is used to weave synthetic carpets. It plays an important role as a reagent in the synthesis of pharmaceutical intermediates and alfa ω-dicarboxylic acid esters. It is a good nucleophile and a michael acceptor to prepare polyamidoamine (PAMAM) dendrimers.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents and peroxides.

Chemical Identifiers

CAS 96-33-3
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008627
InChI Key BAPJBEWLBFYGME-UHFFFAOYSA-N
Synonym methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
PubChem CID 7294
ChEBI CHEBI:82482
IUPAC Name methyl prop-2-enoate
SMILES COC(=O)C=C

Specifications

Melting Point -75°C
Density 0.956
Boiling Point 79°C to 81°C
Flash Point −3°C (26°F)
Odor Stench
Refractive Index 1.403
Quantity 100 mL
UN Number UN1919
Beilstein 605396
Sensitivity Light sensitive
Merck Index 14,6011
Solubility Information Miscible with water.
Formula Weight 86.09
Percent Purity 99%
Chemical Name or Material Methyl acrylate, stabilized with ≈15ppm 4-methoxyphenol
Show More Show Less
Hazard Category H225-H302+H312-H315-H317-H319-H331-H335
Hazard Statement GHS H Statement
H225-H302-H312-H332-H315-H319-H317-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ACRYLATE, STABILIZED
EINECSNumber 202-500-6
RTECSNumber AT2800000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only