missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetylsalicylate 98.0+%, TCI America™

Catalog No. p-7116724
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A011425G 25 g
A0114500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A011425G Supplier TCI America Supplier No. A011425G

Chemical Identifiers

CAS 580-02-9
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00014978
InChI Key ONWPLBKWMAUFGZ-UHFFFAOYSA-N
Synonym 2-Acetoxybenzoic Acid Methyl Ester, Methyl 2-Acetoxybenzoate, Acetylsalicylic Acid Methyl Ester
PubChem CID 68484
IUPAC Name methyl 2-acetyloxybenzoate
SMILES CC(=O)OC1=CC=CC=C1C(=O)OC

Specifications

Melting Point 50°C
Color White
Boiling Point 136°C
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Acetylsalicylate