missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetate, Technical, Approx. 75%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M123020LT

 View more versions of this product

Catalog No. 18611667


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

79-20-9
Steel Can
∼75%
1
20 L
Technical

Chemical Identifiers

79-20-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food use. Read and understand the label and Safety Data Sheet (SDS) prior to use.