missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetate, Technical, Approx. 75%, Spectrum™ Chemical

Catalog No. 18611666
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611666 1 L Amber Glass Bottle
18611669 4 L Amber Glass Bottle
18611667 20 L Steel Can
18611668 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611666 Supplier Spectrum Chemical Mfg Cor Supplier No. M12301LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 79-20-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L
Percent Purity ∼75%
Grade Technical

CAUTION: For manufacturing or laboratory use only. Not for food use. Read and understand the label and Safety Data Sheet (SDS) prior to use.