missing translation for 'onlineSavingsMsg'
Learn More

Methyl acetate, 99+%, Extra Dry, AcroSeal™

Catalog No. p-3593461
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC371831000 100 mL
AC371830010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC371831000 Supplier Thermo Scientific Chemicals Supplier No. 371831000
Only null left

CAS: 79-20-9 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC

Specifications

Melting Point -98.0°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 57.4°C
Flash Point −10°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal glass Bottle
Refractive Index 1.3600 to 1.3620
Linear Formula CH3CO2CH3
Quantity 100 mL
Residue after Evaporation 0.0005 % max.
Beilstein 02, 224
Specific Gravity 0.93
Merck Index 15, 6082
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: miscible with alcohol and ether
Viscosity 0.38 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 99+%
Grade Extra Dry
Physical Form Solution
Chemical Name or Material Methyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-185-2

RUO – Research Use Only