missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-(Hydroxymethyl)nicotinate 97.0+%, TCI America™

Catalog No. p-7133404
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
M2750200MG 200 mg
M27501G 1 g
2 options

Catalog No. M2750200MG

Supplier: TCI America M2750200MG

Only null left

Chemical Identifiers

CAS 56026-36-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00643821
InChI Key QVLONHSXRWCCAE-UHFFFAOYSA-N
Synonym methyl 6-hydroxymethyl nicotinate, 6-hydroxymethyl-nicotinic acid methyl ester, methyl 6-hydroxymethyl pyridine-3-carboxylate, 6-hydroxymethylnicotinic acid methyl ester, 6-hydroxymethyl-nicotinicacidmethylester, methyl 6-hydroxymethylnicotinate, 3-pyridinecarboxylic acid, 6-hydroxymethyl-, methyl ester, 3-pyridinecarboxylicacid, 6-hydroxymethyl-, methyl ester, pubchem19720, methylhydroxymethylnicotinate
PubChem CID 789414
IUPAC Name methyl 6-(hydroxymethyl)pyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)CO

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 200 mg
Formula Weight 167.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-(Hydroxymethyl)nicotinate