missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Hydroxy-2-naphthoate 98.0+%, TCI America™

Catalog No. H10535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10535G 5 g
H105325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10535G Supplier TCI America Supplier No. H10535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17295-11-3
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.21
MDL Number MFCD00100526
InChI Key UKZOPQRTQJERQC-UHFFFAOYSA-N
Synonym 6-Hydroxy-2-naphthoic Acid Methyl Ester
PubChem CID 390995
IUPAC Name methyl 6-hydroxynaphthalene-2-carboxylate
SMILES COC(=O)C1=CC=C2C=C(O)C=CC2=C1

Specifications

Melting Point 170°C
Color White
Quantity 5 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Hydroxy-2-naphthoate