missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Chloronicotinate 98.0+%, TCI America™

Catalog No. p-7132949
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19805G 5 g
M198025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19805G Supplier TCI America Supplier No. M19805G
Only null left

Chemical Identifiers

CAS 73781-91-6
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00023420
InChI Key RMEDXVIWDFLGES-UHFFFAOYSA-N
Synonym methyl 6-chloronicotinate, 6-chloronicotinic acid methyl ester, methyl6-chloronicotinate, methyl 2-chloropyridine-5-carboxylate, 3-pyridinecarboxylic acid, 6-chloro-, methyl ester, methyl 6-chloro-3-pyridinecarboxylate, 6-chloro-3-pyridinecarboxylic acid methyl ester, pubchem13584, methyl-6-chloronicotinate, methyl 6-chloro-nicotinate
PubChem CID 254249
IUPAC Name methyl 6-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)Cl

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 5 g
Formula Weight 171.58
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Chloronicotinate