accessibility menu, dialog, popup

UserName

CAS RN 73781-91-6

IUPAC Name: methyl 6-chloropyridine-3-carboxylate
Synonyms: methyl 6-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: RMEDXVIWDFLGES-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Cl)N=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)