missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromoindole-2-carboxylate, 97%

Catalog No. p-7054778
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6318703 1 g
AAH6318706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6318703 Supplier Thermo Scientific Chemicals Supplier No. H6318703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 372089-59-3 | C11H10BrNO2 | 268.11 g/mol

Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in acetone.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 372089-59-3
MDL Number MFCD03094942
Synonym ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester
PubChem CID 7009496
IUPAC Name ethyl 6-bromo-1H-indole-2-carboxylate

Specifications

Color White
Molecular Formula C11H10BrNO2
Quantity 1 g
Solubility Information Soluble in acetone.
InChI Key FVMZWWFKRMBNSZ-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC2=C(N1)C=C(C=C2)Br
Molecular Weight (g/mol) 268.11
Formula Weight 254.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-bromoindole-2-carboxylate
Show More Show Less
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only