accessibility menu, dialog, popup

UserName

H3COOHNBr

CAS RN 372089-59-3

IUPAC Name: methyl 6-bromo-1H-indole-2-carboxylate
Synonyms: methyl 6-bromo-1H-indole-2-carboxylate
Molecular Weight (g/mol): 254.08
Molecular Formula: C10H8BrNO2
InChi Key: BUMYRKNZGPXFEG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC=C(Br)C=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)