missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 6-bromo-5-fluoronicotinate, 98%
Description
Useful intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Useful intermediate for pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent. Store in freezer at (-20°C).
Useful intermediate for pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent. Store in freezer at (-20°C).
Specifications
Specifications
| Quantity | 1 g |
| Sensitivity | Light sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 234.02 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 6-bromo-5-fluoronicotinate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only