missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromo-5-fluoronicotinate, 98%

Catalog No. AAH6439503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6439503 1 g
AAH6439506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6439503 Supplier Thermo Scientific Chemicals Supplier No. H6439503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1214336-88-5 | C7H5BrFNO2 | 234.02 g/mol

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent. Store in freezer at (-20°C).

Chemical Identifiers

CAS 1214336-88-5
Molecular Formula C7H5BrFNO2
Molecular Weight (g/mol) 234.02
MDL Number MFCD14698110
InChI Key PXHKAVXJQLVUBI-UHFFFAOYSA-N
Synonym methyl 6-bromo-5-fluoronicotinate, 6-bromo-5-fluoro-nicotinic acid methyl ester, 2-bromo-3-fluoro-5-methoxycarbonyl pyridine, methyl6-bromo-5-fluoronicotinate, abbypharma ap-30-7441, 6-bromo-5-fluoropyridine-3-carboxylate, methyl 2-bromo-3-fluoro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-bromo-5-fluoro-, methyl ester
PubChem CID 46311203
IUPAC Name methyl 6-bromo-5-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CC(F)=C(Br)N=C1

Specifications

Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 234.02
Percent Purity 98%
Chemical Name or Material Methyl 6-bromo-5-fluoronicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only