missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 6-Bromo-2-naphthoate 98.0+%, TCI America™
Supplier: TCI America B220225G
Specifications
| Methyl 6-Bromo-2-naphthoate | |
| 125°C | |
| C12H9BrO2 | |
| 25 g | |
| JEUBRLPXJZOGPX-UHFFFAOYSA-N | |
| methyl 6-bromonaphthalene-2-carboxylate | |
| 854134 | |
| ≥98.0% (GC) |
| 33626-98-1 | |
| White-Yellow | |
| MFCD00100408 | |
| methyl 6-bromo-2-naphthoate, methyl6-bromo-2-naphthoate, 6-bromo-2-naphthalenecarboxylic acid methyl ester, 6-bromo-2-naphthoic acid methyl ester, 2-naphthalenecarboxylic acid, 6-bromo-, methyl ester, 6-bromonaphthalene-2-carboxylic acid methyl ester, 6-bromo-naphthalene-2-carboxylic acid methyl ester, pubchem9467, acmc-209i1z, 6-bromo-2-methylnaphthoate | |
| COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br | |
| 265.106 | |
| 265.11 | |
| Crystalline Powder |
Chemical Identifiers
| 33626-98-1 | |
| 265.106 | |
| JEUBRLPXJZOGPX-UHFFFAOYSA-N | |
| 854134 | |
| COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br |
| C12H9BrO2 | |
| MFCD00100408 | |
| methyl 6-bromo-2-naphthoate, methyl6-bromo-2-naphthoate, 6-bromo-2-naphthalenecarboxylic acid methyl ester, 6-bromo-2-naphthoic acid methyl ester, 2-naphthalenecarboxylic acid, 6-bromo-, methyl ester, 6-bromonaphthalene-2-carboxylic acid methyl ester, 6-bromo-naphthalene-2-carboxylic acid methyl ester, pubchem9467, acmc-209i1z, 6-bromo-2-methylnaphthoate | |
| methyl 6-bromonaphthalene-2-carboxylate |
Safety and Handling
TSCA : No