missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Bromo-2-naphthoate 98.0+%, TCI America™

Catalog No. B22025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22025G 5 g
B220225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22025G Supplier TCI America Supplier No. B22025G

Chemical Identifiers

CAS 33626-98-1
Molecular Formula C12H9BrO2
Molecular Weight (g/mol) 265.106
MDL Number MFCD00100408
InChI Key JEUBRLPXJZOGPX-UHFFFAOYSA-N
Synonym methyl 6-bromo-2-naphthoate, methyl6-bromo-2-naphthoate, 6-bromo-2-naphthalenecarboxylic acid methyl ester, 6-bromo-2-naphthoic acid methyl ester, 2-naphthalenecarboxylic acid, 6-bromo-, methyl ester, 6-bromonaphthalene-2-carboxylic acid methyl ester, 6-bromo-naphthalene-2-carboxylic acid methyl ester, pubchem9467, acmc-209i1z, 6-bromo-2-methylnaphthoate
PubChem CID 854134
IUPAC Name methyl 6-bromonaphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 5 g
Formula Weight 265.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Bromo-2-naphthoate