missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 6-bromo-2-naphthoate, 98%
CAS: 33626-98-1 | C12H9BrO2 | 265.1 g/mol
$71.50 - $71.50
Chemical Identifiers
| CAS | 33626-98-1 |
|---|---|
| Molecular Formula | C12H9BrO2 |
| Molecular Weight (g/mol) | 265.1 |
| MDL Number | MFCD00100408 |
| InChI Key | JEUBRLPXJZOGPX-UHFFFAOYSA-N |
| PubChem CID | 854134 |
| IUPAC Name | methyl 6-bromonaphthalene-2-carboxylate |
| SMILES | COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC364140050
|
Thermo Scientific Chemicals
364140050 |
5 g | Glass bottle |
Each for $71.50
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 33626-98-1 | |
| 265.1 | |
| JEUBRLPXJZOGPX-UHFFFAOYSA-N | |
| methyl 6-bromonaphthalene-2-carboxylate |
| C12H9BrO2 | |
| MFCD00100408 | |
| 854134 | |
| COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br |
Specifications
| 33626-98-1 | |
| Brown or Cream to White | |
| 97.5% min. (GC) | |
| C12H9BrO2 | |
| MFCD00100408 | |
| methyl 6-bromo-2-naphthoate, methyl6-bromo-2-naphthoate, 6-bromo-2-naphthalenecarboxylic acid methyl ester, 6-bromo-2-naphthoic acid methyl ester, 2-naphthalenecarboxylic acid, 6-bromo-, methyl ester, 6-bromonaphthalene-2-carboxylic acid methyl ester, 6-bromo-naphthalene-2-carboxylic acid methyl ester, pubchem9467, acmc-209i1z, 6-bromo-2-methylnaphthoate | |
| COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br | |
| 265.1 | |
| 265.1 | |
| Crystalline Powder |
| 123.0°C to 126.0°C | |
| Authentic | |
| Glass bottle | |
| BrC10H6COOCH3 | |
| 5 g | |
| JEUBRLPXJZOGPX-UHFFFAOYSA-N | |
| methyl 6-bromonaphthalene-2-carboxylate | |
| 854134 | |
| 98% | |
| Methyl 6-bromo-2-naphthoate, 98% |
RUO – Research Use Only