missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromo-2-naphthoate, 98%

Catalog No. p-7028002
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2510606 5 g
AAB2510614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2510606 Supplier Thermo Scientific Chemicals Supplier No. B2510606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33626-98-1 | C12H9BrO2 | 265.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33626-98-1
Molecular Formula C12H9BrO2
Molecular Weight (g/mol) 265.106
MDL Number MFCD00100408
InChI Key JEUBRLPXJZOGPX-UHFFFAOYSA-N
Synonym methyl 6-bromo-2-naphthoate, methyl6-bromo-2-naphthoate, 6-bromo-2-naphthalenecarboxylic acid methyl ester, 6-bromo-2-naphthoic acid methyl ester, 2-naphthalenecarboxylic acid, 6-bromo-, methyl ester, 6-bromonaphthalene-2-carboxylic acid methyl ester, 6-bromo-naphthalene-2-carboxylic acid methyl ester, pubchem9467, acmc-209i1z, 6-bromo-2-methylnaphthoate
PubChem CID 854134
IUPAC Name methyl 6-bromonaphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 121°C to 123°C
Quantity 5 g
Beilstein 2578943
Formula Weight 265.11
Percent Purity 98%
Chemical Name or Material Methyl 6-bromo-2-naphthoate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only