missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Aminonicotinate 98.0+%, TCI America™

Catalog No. p-7133029
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21075G 5 g
M210725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21075G Supplier TCI America Supplier No. M21075G
Only null left

Chemical Identifiers

CAS 36052-24-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00797844
InChI Key JACPDLJUQLKABC-UHFFFAOYSA-N
PubChem CID 737487
IUPAC Name methyl 6-aminopyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N

Specifications

Melting Point 160°C
Quantity 5 g
Synonym methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Aminonicotinate