accessibility menu, dialog, popup

UserName

CH3OONH2HN

CAS RN 36052-24-1

IUPAC Name: 2-amino-5-(methoxycarbonyl)pyridin-1-ium
Synonyms: 2-amino-5-(methoxycarbonyl)pyridin-1-ium
Molecular Weight (g/mol): 153.16
Molecular Formula: C7H9N2O2
InChi Key: JACPDLJUQLKABC-UHFFFAOYSA-O
SMILES: COC(=O)C1=CC=C(N)[NH+]=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)