missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-aminonicotinate, 97%

Catalog No. p-7030044
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2757503 1 g
AAH2757506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2757503 Supplier Thermo Scientific Chemicals Supplier No. H2757503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36052-24-1 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36052-24-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00797844
InChI Key JACPDLJUQLKABC-UHFFFAOYSA-N
Synonym methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
PubChem CID 737487
IUPAC Name methyl 6-aminopyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N

Specifications

Melting Point 158°C to 161°C
Quantity 1 g
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Methyl 6-aminonicotinate
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only