missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Amino-2-naphthoate 98.0+%, TCI America™

Catalog No. p-7117558
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16431G 1 g
A16435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16431G Supplier TCI America Supplier No. A16431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5159-59-1
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.225
MDL Number MFCD06797040
InChI Key LPXVPYIHRFOTJZ-UHFFFAOYSA-N
Synonym 6-Amino-2-naphthoic Acid Methyl Ester
PubChem CID 11995029
IUPAC Name methyl 6-aminonaphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)N

Specifications

Melting Point 165°C
Quantity 1 g
Formula Weight 201.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Amino-2-naphthoate