missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Norbornene-2-carboxylate (endo- and exo- mixture) 96.0+%, TCI America™

Catalog No. N069125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N069125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N069125G Supplier TCI America Supplier No. N069125G
Only null left

Chemical Identifiers

CAS 6203-08-3
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00167592
InChI Key RMAZRAQKPTXZNL-UHFFFAOYNA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid Methyl Ester, Methyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, 5-Norbornene-2-carboxylic Acid Methyl Ester
PubChem CID 250075
IUPAC Name methyl bicyclo[2.2.1]hept-2-ene-5-carboxylate
SMILES COC(=O)C1CC2CC1C=C2

Specifications

Color Yellow
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Norbornene-2-carboxylate (endo- and exo- mixture)