missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-nitro-2-furoate, 97%

Catalog No. AAA1522606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1522606 5 g
AAA1522614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1522606 Supplier Thermo Scientific Chemicals Supplier No. A1522606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1874-23-3 | C6H5NO5 | 171.108 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1874-23-3
Molecular Formula C6H5NO5
Molecular Weight (g/mol) 171.108
MDL Number MFCD00051956
InChI Key UTLKCGPAJUYGOM-UHFFFAOYSA-N
Synonym methyl 5-nitro-2-furoate, methyl nitrofuroate, methyl 5-nitrofuroate, 5-nitro-2-furoic acid methyl ester, methyl 5-nitropyromucate, methyl 5-nitro-2-furancarboxylate, unii-71qm4e6j90, ccris 2152, 5-nitro-2-furancarboxylic acid methyl ester, 2-furancarboxylic acid, 5-nitro-, methyl ester
PubChem CID 94882
IUPAC Name methyl 5-nitrofuran-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)[N+](=O)[O-]

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
Beilstein 165132
Formula Weight 171.11
Percent Purity 97%
Chemical Name or Material Methyl 5-nitro-2-furoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
RTECSNumber LV2013000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only