CAS RN 1874-23-3
IUPAC Name:
methyl 5-nitrofuran-2-carboxylate
Synonyms:
methyl 5-nitrofuran-2-carboxylate
Molecular Weight (g/mol):
171.11
Molecular Formula:
C6H5NO5
InChi Key:
UTLKCGPAJUYGOM-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(O1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Methyl 5-nitro-2-furoate, 97%
Methyl 5-nitro-2-furoate, 97%
CAS: 1874-23-3 Molecular Formula: C6H5NO5 Molecular Weight (g/mol): 171.108 MDL Number: MFCD00051956 InChI Key: UTLKCGPAJUYGOM-UHFFFAOYSA-N Synonym: methyl 5-nitro-2-furoate,methyl nitrofuroate,methyl 5-nitrofuroate,5-nitro-2-furoic acid methyl ester,methyl 5-nitropyromucate,methyl 5-nitro-2-furancarboxylate,unii-71qm4e6j90,ccris 2152,5-nitro-2-furancarboxylic acid methyl ester,2-furancarboxylic acid, 5-nitro-, methyl ester PubChem CID: 94882 IUPAC Name: methyl 5-nitrofuran-2-carboxylate SMILES: COC(=O)C1=CC=C(O1)[N+](=O)[O-]