missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Methylsalicylate 96.0+%, TCI America™
$147.25 - $332.17
Chemical Identifiers
| CAS | 22717-57-3 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00016485 |
| InChI Key | JQYUQKRFSSSGJM-UHFFFAOYSA-N |
| Synonym | methyl 5-methylsalicylate, benzoic acid, 2-hydroxy-5-methyl-, methyl ester, 2-hydroxy-5-methylbenzoic acid methyl ester, methyl-5-methylsalicylate, 2,5-cresotic acid, methyl ester, methyl 5-methyl salicylate, p-cresotic acid methyl ester, 5-methylsalicylic acid methyl ester, methyl 2-hydroxy-5-methyl-benzoate, 6-hydroxy-m-toluic acid methyl ester |
| PubChem CID | 89802 |
| IUPAC Name | methyl 2-hydroxy-5-methylbenzoate |
| SMILES | CC1=CC(=C(C=C1)O)C(=O)OC |
Chemical Identifiers
| 22717-57-3 | |
| 166.176 | |
| JQYUQKRFSSSGJM-UHFFFAOYSA-N | |
| 89802 | |
| CC1=CC(=C(C=C1)O)C(=O)OC |
| C9H10O3 | |
| MFCD00016485 | |
| methyl 5-methylsalicylate, benzoic acid, 2-hydroxy-5-methyl-, methyl ester, 2-hydroxy-5-methylbenzoic acid methyl ester, methyl-5-methylsalicylate, 2,5-cresotic acid, methyl ester, methyl 5-methyl salicylate, p-cresotic acid methyl ester, 5-methylsalicylic acid methyl ester, methyl 2-hydroxy-5-methyl-benzoate, 6-hydroxy-m-toluic acid methyl ester | |
| methyl 2-hydroxy-5-methylbenzoate |
Specifications
| 22717-57-3 | |
| 242°C | |
| MFCD00016485 | |
| methyl 5-methylsalicylate, benzoic acid, 2-hydroxy-5-methyl-, methyl ester, 2-hydroxy-5-methylbenzoic acid methyl ester, methyl-5-methylsalicylate, 2,5-cresotic acid, methyl ester, methyl 5-methyl salicylate, p-cresotic acid methyl ester, 5-methylsalicylic acid methyl ester, methyl 2-hydroxy-5-methyl-benzoate, 6-hydroxy-m-toluic acid methyl ester | |
| CC1=CC(=C(C=C1)O)C(=O)OC | |
| 166.176 | |
| 166.18 | |
| Liquid |
| Yellow | |
| C9H10O3 | |
| 5 g | |
| JQYUQKRFSSSGJM-UHFFFAOYSA-N | |
| methyl 2-hydroxy-5-methylbenzoate | |
| 89802 | |
| ≥96.0% (GC) | |
| Methyl 5-Methylsalicylate |
Safety and Handling
TSCA : Yes