missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Methylsalicylate 96.0+%, TCI America™

Catalog No. M13535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13535G 5 g
M135325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13535G Supplier TCI America Supplier No. M13535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22717-57-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016485
InChI Key JQYUQKRFSSSGJM-UHFFFAOYSA-N
Synonym methyl 5-methylsalicylate, benzoic acid, 2-hydroxy-5-methyl-, methyl ester, 2-hydroxy-5-methylbenzoic acid methyl ester, methyl-5-methylsalicylate, 2,5-cresotic acid, methyl ester, methyl 5-methyl salicylate, p-cresotic acid methyl ester, 5-methylsalicylic acid methyl ester, methyl 2-hydroxy-5-methyl-benzoate, 6-hydroxy-m-toluic acid methyl ester
PubChem CID 89802
IUPAC Name methyl 2-hydroxy-5-methylbenzoate
SMILES CC1=CC(=C(C=C1)O)C(=O)OC

Specifications

Color Yellow
Boiling Point 242°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Methylsalicylate