missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Methylpyrazole-3-carboxylate 98.0+%, TCI America™

Catalog No. p-7133234
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24651G 1 g
M24655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24651G Supplier TCI America Supplier No. M24651G
Only null left

Chemical Identifiers

CAS 25016-17-5
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD03778987
InChI Key GFEZEVUIYRGWNU-UHFFFAOYSA-N
Synonym methyl 3-methyl-1h-pyrazole-5-carboxylate, methyl 5-methylpyrazole-3-carboxylate, methyl 5-methyl-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-methyl-, methyl ester, 5-methyl-1h-pyrazole-3-carboxylic acid methyl ester, 5-methyl-2h-pyrazole-3-carboxylic acid methyl ester, 5-methyl-1 2 h-pyrazole-3-carboxylic acid methyl ester, methylmethylpyrazolecarboxylate, ksc554q1l, 5-methoxycarbonyl-3-me-thylpyrazole
PubChem CID 2753079
IUPAC Name methyl 5-methyl-1H-pyrazole-3-carboxylate
SMILES CC1=CC(=NN1)C(=O)OC

Specifications

Melting Point 83°C
Color White
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Methylpyrazole-3-carboxylate