missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Methoxy-3-oxovalerate 96.0+%, TCI America™
$398.90 - $1590.03
Chemical Identifiers
| CAS | 62462-05-9 |
|---|---|
| Molecular Formula | C7H12O4 |
| Molecular Weight (g/mol) | 160.17 |
| MDL Number | MFCD00042840 |
| InChI Key | OPURNNHYTATPKM-UHFFFAOYSA-N |
| Synonym | 3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate |
| PubChem CID | 542416 |
| IUPAC Name | methyl 5-methoxy-3-oxopentanoate |
| SMILES | COCCC(=O)CC(=O)OC |
Chemical Identifiers
| 62462-05-9 | |
| 160.17 | |
| OPURNNHYTATPKM-UHFFFAOYSA-N | |
| 542416 | |
| COCCC(=O)CC(=O)OC |
| C7H12O4 | |
| MFCD00042840 | |
| 3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate | |
| methyl 5-methoxy-3-oxopentanoate |
Specifications
| 62462-05-9 | |
| 69°C | |
| MFCD00042840 | |
| 3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate | |
| COCCC(=O)CC(=O)OC | |
| 160.17 | |
| 160.17 | |
| Liquid |
| Yellow | |
| C7H12O4 | |
| 1 g | |
| OPURNNHYTATPKM-UHFFFAOYSA-N | |
| methyl 5-methoxy-3-oxopentanoate | |
| 542416 | |
| ≥96.0% (GC) | |
| Methyl 5-Methoxy-3-oxovalerate |
Safety and Handling
TSCA : No