missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-(Hydroxymethyl)-1H-imidazole-4-carboxylate 98.0+%, TCI America™

Catalog No. H08401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08401G 1 g
H08405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08401G Supplier TCI America Supplier No. H08401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82032-43-7
Molecular Formula C6H8N2O3
Molecular Weight (g/mol) 156.141
MDL Number MFCD00191520
InChI Key RRNBWPNDHRAYBV-UHFFFAOYSA-N
Synonym 5-(Hydroxymethyl)-1H-imidazole-4-carboxylic Acid Methyl Ester
PubChem CID 466070
IUPAC Name methyl 5-(hydroxymethyl)-1H-imidazole-4-carboxylate
SMILES COC(=O)C1=C(NC=N1)CO

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 156.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-(Hydroxymethyl)-1H-imidazole-4-carboxylate