accessibility menu, dialog, popup

UserName

CAS RN 82032-43-7

IUPAC Name: methyl 5-(hydroxymethyl)-1H-imidazole-4-carboxylate
Synonyms: methyl 5-(hydroxymethyl)-1H-imidazole-4-carboxylate
Molecular Weight (g/mol): 156.14
Molecular Formula: C6H8N2O3
InChi Key: RRNBWPNDHRAYBV-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(CO)NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results